Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EU360796.1 | - | 8 - 9 | 1084 | 77 | |
EU812236.1 | - | 8 - 9 | 1084 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Vitis vinifera Probe spans exons |