Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EU925809.1 | - | 1 - 2 | 79 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Vitis vinifera Probe spans exons |