Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhrs9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001191.1 | NP_001001191.1 | 2 - 3 | 391 | 75 | |
XM_012970271.2 | XP_012825725.1 | 2 - 3 | 438 | 75 | ||
GenBank mRNA | BC067304.1 | - | 2 - 3 | 391 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |