Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csnk1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001221.2 | NP_001001221.1 | 9 - 10 | 1309 | 56 | |
XM_012958782.2 | XP_012814236.1 | 10 - 11 | 1539 | 56 | ||
GenBank mRNA | BC067926.2 | - | 9 - 10 | 1309 | 56 | |
CT025205.2 | - | 9 - 10 | 1233 | 56 | ||
CT030065.2 | - | 9 - 10 | 1291 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |