Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc31a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001238.1 | NP_001001238.1 | 446 | 99 | ||
GenBank mRNA | BC067952.1 | - | 446 | 99 | ||
CR761086.2 | - | 424 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis |