Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fut11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002904.2 | NP_001002904.1 | 1 - 2 | 1052 | 77 | |
GenBank mRNA | AJ784816.1 | - | 1 - 2 | 1096 | 77 | |
BC167262.1 | - | 1 - 2 | 905 | 77 | ||
BC171316.1 | - | 1 - 2 | 1016 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |