Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003660.1 | NP_001003660.1 | 595 | 85 | ||
GenBank mRNA | AY204560.1 | - | 595 | 85 |
Gene Symbol: LOC101731954 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004920660.2 | XP_004920717.2 | 3 - 4 | 526 | 85 | |
GenBank mRNA | AY204561.1 | - | 3 - 4 | 526 | 85 | |
BC158547.1 | - | 623 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |