Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004791.1 | NP_001004791.1 | 3 - 4 | 372 | 63 | |
XM_012956698.2 | XP_012812152.1 | 3 - 4 | 493 | 63 | ||
GenBank mRNA | BC074526.1 | - | 3 - 4 | 372 | 63 | |
CR762157.2 | - | 3 - 4 | 343 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |