Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stoml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004808.1 | NP_001004808.1 | 405 | 77 | ||
GenBank mRNA | BC074573.1 | - | 405 | 77 | ||
CR761170.2 | - | 397 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis |