Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pelo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004823.1 | NP_001004823.1 | 916 | 78 | ||
GenBank mRNA | BC074604.1 | - | 916 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis |