Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ran | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004829.1 | NP_001004829.1 | 5 - 6 | 683 | 93 | |
XM_012959006.2 | XP_012814460.1 | 5 - 6 | 485 | 93 | ||
XM_012959012.2 | XP_012814466.1 | 5 - 6 | 633 | 93 | ||
XM_018091941.1 | XP_017947430.1 | 5 - 6 | 584 | 93 | ||
GenBank mRNA | BC074619.1 | - | 5 - 6 | 683 | 93 | |
CR761555.2 | - | 4 - 5 | 448 | 93 | ||
CR761725.2 | - | 4 - 5 | 456 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |