Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgam5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004858.1 | NP_001004858.1 | 2 - 3 | 375 | 63 | |
XM_012959055.2 | XP_012814509.1 | 2 - 3 | 503 | 63 | ||
GenBank mRNA | BC074682.1 | - | 2 - 3 | 375 | 63 | |
CT010555.2 | - | 2 - 3 | 363 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |