Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgam5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004858.1 | NP_001004858.1 | 4 - 5 | 584 | 96 | |
GenBank mRNA | BC074682.1 | - | 4 - 5 | 584 | 96 | |
CT010555.2 | - | 4 - 5 | 572 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |