Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nectin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004860.1 | NP_001004860.1 | 3 - 4 | 812 | 102 | |
XM_012956717.2 | XP_012812171.1 | 3 - 4 | 861 | 102 | ||
GenBank mRNA | BC074691.1 | - | 3 - 4 | 812 | 102 | |
CR762185.2 | - | 3 - 4 | 811 | 102 |
Gene Symbol: LOC108645444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018090656.1 | XP_017946145.1 | 2 - 3 | 631 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |