Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: n4bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004870.1 | NP_001004870.1 | 3 - 4 | 1780 | 67 | |
XM_012960812.2 | XP_012816266.1 | 3 - 4 | 1930 | 67 | ||
GenBank mRNA | BC075104.1 | - | 3 - 4 | 1780 | 67 | |
CT025272.2 | - | 3 - 4 | 1756 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |