Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004871.1 | NP_001004871.1 | 4 - 5 | 335 | 101 | |
GenBank mRNA | BC075111.1 | - | 4 - 5 | 335 | 101 | |
CR855554.2 | - | 3 - 4 | 312 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |