Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmgb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004888.1 | NP_001004888.1 | 4 - 5 | 533 | 88 | |
XM_018096198.1 | XP_017951687.1 | 4 - 5 | 570 | 88 | ||
GenBank mRNA | BC075290.1 | - | 4 - 5 | 533 | 88 | |
CR761259.2 | - | 4 - 5 | 562 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |