Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004893.1 | NP_001004893.1 | 4 - 5 | 350 | 93 | |
XM_012956730.2 | XP_012812184.1 | 4 - 5 | 361 | 93 | ||
XM_018091159.1 | XP_017946648.1 | 4 - 6 | 96 | |||
XM_018091160.1 | XP_017946649.1 | 4 - 6 | 96 | |||
GenBank mRNA | BC075297.1 | - | 4 - 5 | 350 | 93 | |
CR761375.2 | - | 4 - 5 | 289 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |