Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mitd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004902.1 | NP_001004902.1 | 2 - 3 | 310 | 96 | |
GenBank mRNA | BC075321.1 | - | 2 - 3 | 310 | 96 | |
CR761296.2 | - | 2 - 3 | 299 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |