Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mcee | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004927.1 | NP_001004927.1 | 1 - 2 | 71 | 57 | |
GenBank mRNA | BC075386.1 | - | 1 - 2 | 71 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |