Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sav1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004931.1 | NP_001004931.1 | 1 - 2 | 253 | 69 | |
GenBank mRNA | BC075399.1 | - | 1 - 2 | 253 | 69 | |
CR760942.2 | - | 1 - 2 | 104 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |