Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bicdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004934.1 | NP_001004934.1 | 530 | 71 | ||
GenBank mRNA | BC075409.1 | - | 530 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis |