Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pinx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004938.1 | NP_001004938.1 | 3 - 4 | 281 | 101 | |
XM_012964033.2 | XP_012819487.1 | 3 - 4 | 324 | 101 | ||
GenBank mRNA | BC075416.1 | - | 3 - 4 | 281 | 101 | |
EU520259.1 | - | 3 - 4 | 228 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |