Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001247979.1 | NP_001234908.1 | 5 - 6 | 634 | 109 | |
NM_001247980.1 | NP_001234909.1 | 5 - 6 | 598 | 109 | ||
NR_045263.1 | 5 - 6 | 910 | 109 | |||
XM_018092025.1 | XP_017947514.1 | 6 - 7 | 801 | 109 | ||
XM_018092026.1 | XP_017947515.1 | 5 - 6 | 731 | 109 | ||
XM_018092027.1 | XP_017947516.1 | 5 - 6 | 724 | 109 | ||
GenBank mRNA | BC075421.1 | - | 5 - 6 | 902 | 109 | |
CX935094.1 | - | 5 - 6 | 580 | 109 | ||
EL815463.1 | - | 5 - 6 | 623 | 109 | ||
EL818546.1 | - | 5 - 6 | 621 | 109 | ||
EL827396.1 | - | 5 - 6 | 603 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |