Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abitram | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004966.1 | NP_001004966.1 | 6 - 7 | 621 | 104 | |
XM_012964525.2 | XP_012819979.1 | 7 - 8 | 756 | 104 | ||
XM_012964526.2 | XP_012819980.1 | 6 - 7 | 590 | 104 | ||
XM_012964527.2 | XP_012819981.1 | 6 - 7 | 587 | 104 | ||
GenBank mRNA | BC075478.1 | - | 6 - 7 | 621 | 104 | |
CR761531.2 | - | 6 - 7 | 502 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |