Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005006.1 | NP_001005006.1 | 3 - 4 | 1368 | 71 | |
XM_012959921.2 | XP_012815375.1 | 3 - 4 | 1315 | 71 | ||
GenBank mRNA | BC076652.1 | - | 3 - 4 | 1368 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |