Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c19orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005016.1 | NP_001005016.1 | 5 - 6 | 415 | 63 | |
XM_018096214.1 | XP_017951703.1 | 5 - 6 | 587 | 63 | ||
GenBank mRNA | BC076686.1 | - | 5 - 6 | 415 | 63 | |
CR848118.2 | - | 5 - 6 | 360 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |