Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsph9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005021.1 | NP_001005021.1 | 470 | 99 | ||
GenBank mRNA | BC076699.1 | - | 470 | 99 | ||
CR760050.2 | - | 397 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis |