Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005027.3 | NP_001005027.3 | 7 - 8 | 1422 | 88 | |
XM_012959973.2 | XP_012815427.1 | 7 - 8 | 1719 | 88 | ||
XM_018092265.1 | XP_017947754.1 | 7 - 8 | 1686 | 88 | ||
XM_018092272.1 | XP_017947761.1 | 7 - 8 | 1746 | 88 | ||
XM_018092275.1 | XP_017947764.1 | 7 - 8 | 1441 | 88 | ||
GenBank mRNA | AL781524.2 | - | 1 - 2 | 139 | 88 | |
BC076709.1 | - | 7 - 8 | 1411 | 88 | ||
BX687753.1 | - | 1 - 2 | 469 | 88 | ||
CF218052.1 | - | 3 - 4 | 704 | 88 | ||
CU025035.1 | - | 7 - 8 | 1391 | 88 | ||
CX449218.1 | - | 1 - 2 | 695 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |