Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005041.1 | NP_001005041.1 | 2 - 3 | 282 | 126 | |
XM_012968241.2 | XP_012823695.1 | 2 - 3 | 260 | 126 | ||
XM_012968243.2 | XP_012823697.1 | 2 - 3 | 308 | 126 | ||
XM_012968244.2 | XP_012823698.1 | 2 - 3 | 298 | 126 | ||
XM_012968245.2 | XP_012823699.1 | 2 - 3 | 267 | 126 | ||
XM_012968246.2 | XP_012823700.1 | 2 - 3 | 192 | 126 | ||
GenBank mRNA | BC076904.1 | - | 2 - 3 | 282 | 126 | |
CR761196.2 | - | 1 - 2 | 173 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |