Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005055.2 | NP_001005055.2 | 6 - 7 | 1132 | 75 | |
XM_018091171.1 | XP_017946660.1 | 5 - 6 | 1047 | 75 | ||
GenBank mRNA | BC076943.1 | - | 6 - 7 | 1132 | 75 | |
CU075748.1 | - | 6 - 7 | 793 | 75 | ||
CX384657.2 | - | 3 - 4 | 483 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |