Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: otud3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005056.1 | NP_001005056.1 | 2 - 3 | 372 | 64 | |
GenBank mRNA | BC076944.1 | - | 3 - 4 | 402 | 64 | |
CR760548.2 | - | 2 - 3 | 390 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |