Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tada1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005070.1 | NP_001005070.1 | 8 - 9 | 968 | 92 | |
XM_018093002.1 | XP_017948491.1 | 8 - 9 | 941 | 92 | ||
GenBank mRNA | BC076979.1 | - | 8 - 9 | 968 | 92 | |
CR760219.2 | - | 8 - 9 | 1057 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |