Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005071.1 | NP_001005071.1 | 5 - 6 | 1133 | 81 | |
XM_012960880.2 | XP_012816334.1 | 6 - 7 | 876 | 81 | ||
XM_018093003.1 | XP_017948492.1 | 6 - 7 | 890 | 81 | ||
XM_018093004.1 | XP_017948493.1 | 6 - 7 | 987 | 81 | ||
XM_018093005.1 | XP_017948494.1 | 6 - 7 | 1190 | 81 | ||
XM_018093006.1 | XP_017948495.1 | 6 - 7 | 848 | 81 | ||
GenBank mRNA | BC076980.1 | - | 5 - 6 | 1133 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |