Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005073.1 | NP_001005073.1 | 6 - 7 | 696 | 92 | |
XM_012968255.2 | XP_012823709.1 | 6 - 7 | 813 | 92 | ||
GenBank mRNA | BC076990.1 | - | 6 - 7 | 696 | 92 | |
CU075836.1 | - | 6 - 7 | 683 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |