Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tvp23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005091.1 | NP_001005091.1 | 4 - 5 | 369 | 68 | |
GenBank mRNA | BC077019.1 | - | 4 - 5 | 369 | 68 | |
CT025310.2 | - | 3 - 4 | 507 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |