Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pomp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005093.2 | NP_001005093.1 | 4 - 5 | 357 | 79 | |
GenBank mRNA | BC077023.1 | - | 4 - 5 | 359 | 79 | |
CR855731.2 | - | 4 - 5 | 307 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |