Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnaja1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011012.1 | NP_001011012.1 | 4 - 5 | 546 | 82 | |
GenBank mRNA | BC082725.1 | - | 4 - 5 | 546 | 82 | |
CT025404.2 | - | 4 - 5 | 484 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |