Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203523.1 | NP_988854.1 | 5 - 6 | 739 | 81 | |
GenBank mRNA | AY079192.1 | - | 5 - 6 | 739 | 81 | |
CR926385.2 | - | 5 - 6 | 728 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Xenopus tropicalis Probe spans exons |