Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203523.1 | NP_988854.1 | 7 - 8 | 973 | 98 | |
GenBank mRNA | AY079192.1 | - | 7 - 8 | 973 | 98 | |
CR926385.2 | - | 7 - 8 | 962 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Xenopus tropicalis Probe spans exons |