Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ing5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203536.1 | NP_988867.1 | 5 - 6 | 537 | 92 | |
GenBank mRNA | BC059768.1 | - | 5 - 6 | 537 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |