Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203552.1 | NP_988883.1 | 1 - 2 | 112 | 93 | |
XM_012958741.2 | XP_012814195.1 | 1 - 2 | 135 | 93 | ||
GenBank mRNA | BC059736.1 | - | 1 - 2 | 112 | 93 | |
CR761659.1 | - | 1 - 2 | 75 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |