Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: atp6v0c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203562.1 | NP_988893.1 | 1 - 2 | 226 | 75 | |
GenBank mRNA | BC059745.1 | - | 1 - 2 | 226 | 75 | |
CT030632.1 | - | 1 - 2 | 89 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |