Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203618.1 | NP_988949.1 | 1 - 2 | 623 | 98 | |
GenBank mRNA | BC061326.1 | - | 1 - 2 | 623 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |