Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nkx2-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203620.1 | NP_988951.1 | 1 - 2 | 276 | 60 | |
GenBank mRNA | BC061321.1 | - | 1 - 2 | 276 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |