Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203634.1 | NP_988965.1 | 3 - 4 | 455 | 61 | |
GenBank mRNA | BC061429.1 | - | 3 - 4 | 455 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |