Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203642.1 | NP_988973.1 | 2 - 3 | 431 | 95 | |
GenBank mRNA | BC061386.1 | - | 2 - 3 | 431 | 95 | |
CR848567.2 | - | 2 - 3 | 223 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |