Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203658.1 | NP_988989.1 | 1 - 2 | 208 | 78 | |
GenBank mRNA | BC061305.1 | - | 1 - 2 | 208 | 78 | |
CR848395.2 | - | 1 - 2 | 153 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |