Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203662.1 | NP_988993.1 | 3 - 4 | 351 | 85 | |
GenBank mRNA | BC061284.1 | - | 3 - 4 | 351 | 85 | |
CR760776.2 | - | 3 - 4 | 356 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |