Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203710.1 | NP_989041.1 | 3 - 4 | 391 | 76 | |
XM_012968151.2 | XP_012823605.1 | 2 - 3 | 528 | 76 | ||
XM_012968152.2 | XP_012823606.1 | 2 - 3 | 531 | 76 | ||
GenBank mRNA | BC061372.1 | - | 3 - 4 | 391 | 76 | |
CR762001.2 | - | 2 - 3 | 185 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |